missing translation for 'onlineSavingsMsg'
Learn More

TCR V alpha 11, APC, clone: RR8-1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5800-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15320760

  • £69.05 / 25µg


Estimated Shipment
05-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This RR8-1 monoclonal antibody reacts with the mouse T cell receptor (TCR) V alpha 11 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The RR8-1 antibody recognizes the V beta 11 chain on T cells from mouse strains with the b (e.g., C57BL/6, B10) and d (e.g., DBA/1, DBA/2) haplotypes. V alpha 11+ T cells are absent in strains with the a (e.g., BALB/c, C3H) or c (e.g., SJL) haplotypes of the Tcra gene complex. Finally, V alpha 11 is expressed predominantly on CD4+ T cells.The RR8-1 antibody recognizes the V alpha 11.1 and 11.2 alleles, but not 11.3. This antibody has been reported to stimulate V alpha 11+ T cells.Applications Reported: This RR8-1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This RR8-1 antibody has been tested by flow cytometric analysis of mouse lymph node. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

V alpha 11 TCR
RR8-1
APC
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Valpha11, Va11
IgG2b
RUO
Primary
Murine
Videos