Learn More
UVP Filter Glass: Shortwave UV Grid Replacement
Lamps have grid design, which is a highly uniform 254nm UV source of high-intensity
Brand:  UVP 77-0002-02
Code : 03
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 12873029
  • £319.26 / Each
Estimated Shipment
06-07-2018
Add to basket
Description
Description
  • Lamp features a high intensity 254nm UV grid
  • UVP's special UVG shortwave filter reduces solarization, prolonging the filter life
  • Lamp mounts securely in its transformer base or is removable for movement around your work area
  • Lamp manufactured with rugged, scratch-resistant plastic
  • A snap-on filter assembly can be easily attached to convert this lamp for fluorescent analysis
Bacteria destruction, Sterilization, Photochemistry/Phosphorescence, Lapidary, Fluorescent analysis
Specifications
Specifications
Model Mineralight™ R-52G high intensity UV grid lamp
Each
Shortwave Grid
Videos