missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UVP Filter Assemblies for White/UV Light Transilluminators
£317.00 - £710.00
Specifications
Product Type | Filter Assembly |
---|---|
Filtration Area (English) | 7.87 x 7.87 in. |
Filtration Area (Metric) | 20 x 20 cm |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
12823009
|
UVP
38-0189-05 |
TMW-20 UV/white light transilluminator |
£710.00
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
12833009
|
UVP
38-0189-08 |
TLW-20 UV/white light transilluminator |
£317.00
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
Specifications
Filter Assembly | |
7.87 x 7.87 in. | |
20 x 20 cm |