missing translation for 'onlineSavingsMsg'
Learn More

UVI 3003, Tocris Bioscience™

RXR antagonist

Brand:  Tocris (Bio-Techne) 330310

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13462744

  • £170.28 / 10mg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

RXR antagonist; displays high RXR binding affinity. Does not affect the corepressor interaction capacity of the RARα subunit within the context of the RAR-RXR heterodimer.
Specifications

Chemical Identifiers

847239-17-2
436.592
compound 10e pmid: 19408900, d09ien, 3-4-hydroxy-3-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-pentyloxy-2-naphthalenyl phenyl-2-propenoicacid, e-3-4-hydroxy-3-1,1,4,4-tetramethyl-7-pentoxy-tetralin-6-yl phenyl prop-2-enoic acid, 2e-3-4-hydroxy-3-5,5,8,8-tetramethyl-3-pentyloxy-5,6,7,8-tetrahydronaphthalen-2-yl phenyl prop-2-enoic acid, 3-4-hydroxy-3-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-pentyloxy-2-naphthalenyl phenyl-2-propenoic acid
(E)-3-[4-hydroxy-3-(5,5,8,8-tetramethyl-3-pentoxy-6,7-dihydronaphthalen-2-yl)phenyl]prop-2-enoic acid
C28H36O4
APJSHECCIRQQDV-ZRDIBKRKSA-N
44566108
CCCCCOC1=CC2=C(C=C1C3=C(C=CC(=C3)C=CC(=O)O)O)C(CCC2(C)C)(C)C

Specifications

UVI 3003
847239-17-2
compound 10e pmid: 19408900, d09ien, 3-4-hydroxy-3-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-pentyloxy-2-naphthalenyl phenyl-2-propenoicacid, e-3-4-hydroxy-3-1,1,4,4-tetramethyl-7-pentoxy-tetralin-6-yl phenyl prop-2-enoic acid, 2e-3-4-hydroxy-3-5,5,8,8-tetramethyl-3-pentyloxy-5,6,7,8-tetrahydronaphthalen-2-yl phenyl prop-2-enoic acid, 3-4-hydroxy-3-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-pentyloxy-2-naphthalenyl phenyl-2-propenoic acid
CCCCCOC1=CC2=C(C=C1C3=C(C=CC(=C3)C=CC(=O)O)O)C(CCC2(C)C)(C)C
436.592
436.58
10mg
C28H36O4
APJSHECCIRQQDV-ZRDIBKRKSA-N
(E)-3-[4-hydroxy-3-(5,5,8,8-tetramethyl-3-pentoxy-6,7-dihydronaphthalen-2-yl)phenyl]prop-2-enoic acid
44566108
>98%
Videos