Learn More
Veolia Water Systems™ UV Lamp
Rich, robust UV lamp that features UV light acting as a bactericide to break down and photo-oxidize organic contaminants to polar or ionized species.
Brand:  Veolia Water Systems™ LC210-02
Code : 03
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 13135980
  • £916.90 / Each
Estimated Shipment: 03-05-2024
to see stock.
Add to basket
Description
Description
  • UV light is used as a bactericide and to break down and photo-oxidise organic contaminants to polar or ionized species
  • The UV bactericidal action damages DNA and RNA polymerase at low doses thereby preventing replication
Specifications
Specifications
Used as a bactericide and to break down and photo-oxidize organic contaminats to polar or ionized species.
Used as a bactericide and to break down and photo-oxidize organic contaminants to polar or ionized species