missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ UV Cabinets

Brand:  Grant Instruments™ UVC/T-M-AR UK

Code : 04

Additional Details : Weight : 34.70000kg

Product Code. 12915521

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

515 x 690 x 555mm
230V
Internal Working Area: 650 x 475mm
Videos
Documentation