missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ uTIP™ Filter Sterile Low Retention Universal Pipette Tips, Standard

Product Code. 30216537
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
100 μL
200 μL
300 μL
1000 μL
1250 μL
Unit Size:
Pack of 960
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
30216537 200 μL Pack of 960
30216541 10 μL Pack of 960
30216549 20 μL Pack of 960
30216530 100 μL Pack of 960
30216546 300 μL Pack of 960
30216533 1250 μL Pack of 960
30216567 1000 μL Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 30216537 Supplier BIOTIX™ Supplier No. 63300044
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Experience a better universal tip. Biotix™ uTIP™ Filter Sterile Pipette Tips made with Flexfit™ creates a secure seal on various pipettes with less insertion and ejection force.

Experience a better universal tip with Biotix™ uTIP™

  • Biotix filter sterile tips offers aerosol protection to prevent cross contamination
  • Fits all standard universal shaft single channel, multichannel and electronic pipettes
  • FlexFit technology features flexible proximal end to ensure secure seal with 66 to 80% less insertion force than competitor tips
  • X-Resin drastically reduces sample retention resulting in tighter CV values
  • Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
  • Each lot of Biotix tips are tested and certified free of RNase, DNase, Endotoxins/Pyrogens, PCR Inhibitors, trace metals, and human DNA

Certifications

ISO 9001

TRUSTED_SUSTAINABILITY

Specifications

Description 200 μL, Racked, filtered
Volume (Metric) 200 μL
Filtered Filtered
Sterility Sterile
Color Clear
Rack Color Green
Autoclavable Not Autoclavable
Model Universal
Length (Metric) 59.54 mm
Certifications/Compliance ISO 9001
Clean Claims Reusable as a 96 well reservoir
For Use With (Application) Low binding for Protein, DNA, and nanoparticles
Sterilization Method e-beam
Format Hinged Rack
Tip Style Ergonomic
Packaging Racked
Quantity 10 racks/pk
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line Biotix uTip
Material Polypropylene
Shape Conical
Graduations 10 μL, 50 μL, 100 μL, 200 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.