Learn More
UTF1 Monoclonal Antibody (MFCDA84), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-9849-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Undifferentiated transcription factor-1 (UTF-1) is used as marker for the undifferentiated state of pluripotent stem cells.UTF1 is a chromatin-associated protein with core histone-like properties. UTF1 further acts as a transcriptional repressor and is required for proper differentiation of pluripotent cells.
Undifferentiated transcription factor-1 (UTF-1) is used as marker for the undifferentiated state of pluripotent stem cells.UTF1 is a chromatin-associated protein with core histone-like properties. UTF1 further acts as a transcriptional repressor and is required for proper differentiation of pluripotent cells.Specifications
UTF1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q5T230, Q6J1H4 | |
UTF1 | |
Bacterial expression of aa 44-153 (homology between mouse and human 81%) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
MFCDA84 | |
Unconjugated | |
UTF1 | |
AI505934; OTTHUMP00000020786; Undifferentiated embryonic cell transcription factor 1; UTF1 | |
Mouse | |
Affinity chromatography | |
RUO | |
22286, 8433 | |
4° C | |
Liquid |
For Research Use Only.