missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ USP7 Recombinant Human Protein, N-His.GST Tag

Expressed from DNA sequence encoding amino acid sequence of human USP7 fused to N-terminal polyhistidine-tagged GST tag at N-terminus

Brand:  Sino Biological™ 11681H20B50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15288825

  • £78.60 / 50µg


Estimated Shipment
16-05-2018
Add to basket

Description

Description

  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, 0.5mM PMSF, 0.5mM EDTA, 0.5mM GSH, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Lyophilized
68.6kd
50μg
Truncated, Recombinant
Ubiquitin Carboxyl-Terminal Hydrolase 7
Assay Standard
Insect
GST Tag, His Tag
7874
≥95% by SDS-PAGE
Human
<1 EU/μg
USP7
Proteases, Peptidases and Inhibitors
USP7 Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.