Learn More
Thermo Scientific™ USA Standard Sieves, 50 Mesh
Brass frame and Stainless-steel mesh sieves
POA
Specifications
Type Sieve
Material Stainless Steel
Certifications/Compliance ASTM E11
Description 50 Mesh
View More Specs
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Brass frame, stainless steel mesh sieves meet ASTM E11 requirements and all current amendments
  • Sieves are full height
Sieve Depths
  • 3 in. diameter sieves are 1 in. deep
  • 8 in. diameter sieves are 2 in. deep
  • 12 in.diameter sieves are 3 in. deep
Specifications
Specifications
Sieve
Stainless Steel
ASTM E11
50 Mesh