missing translation for 'onlineSavingsMsg'
Learn More

Ursodeoxycholic acid, 99%, ACROS Organics™

Brand:  Acros Organics 208590250

Code : 29

Additional Details : CAS Number : 128-13-2 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10259273

  • £180.60 / 25g


Estimated Shipment
09-02-2018
Add to basket

Specifications

Chemical Identifiers

Ursodeoxycholic acid
≥98.5% (Acid)
C24H40O4
MFCD00003680
128-13-2
99%
392.57

Specifications

Ursodeoxycholic acid
Solution
99%
C24H40O4
MFCD00003680
Glass bottle
+60.00 (20.00°C c=2,C2H5OH)
200° to 206°C
128-13-2
≥98.5% (Acid)
Authentic
392.57
15, 10074
+58° to +62° (20°C, 589nm) (c=2, ethanol)
Clear colorless
25g
Videos
SDS