missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ursodeoxycholic acid, 99%, ACROS Organics™
£18.28 - £188.19
Chemical Identifiers
CAS | 128-13-2 |
---|---|
Molecular Formula | C24H40O4 |
Molecular Weight (g/mol) | 392.58 |
MDL Number | MFCD00003680 |
InChI Key | RUDATBOHQWOJDD-UZVSRGJWSA-N |
Synonym | ursodeoxycholic acid, ursodiol, actigall, ursofalk, ursolvan, delursan, ursodeoxycholate, urso, urso forte, ursodesoxycholic acid |
PubChem CID | 31401 |
ChEBI | CHEBI:9907 |
IUPAC Name | (4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid |
SMILES | CC(CCC(=O)O)C1CCC2C1(CCC3C2C(CC4C3(CCC(C4)O)C)O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10052660
|
Acros Organics
208590010 |
1g | Glass bottle |
£18.28
1g |
In stock, ready to ship: 3 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
10431251
|
Acros Organics
208590050 |
5g | Glass bottle |
£54.43
5g |
In stock, ready to ship: 1 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
10259273
|
Acros Organics
208590250 |
25g | Glass bottle |
£188.19
25g |
Estimated Shipment 06-05-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Chemical Identifiers
128-13-2 | |
392.58 | |
RUDATBOHQWOJDD-UZVSRGJWSA-N | |
31401 | |
(4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid |
C24H40O4 | |
MFCD00003680 | |
ursodeoxycholic acid, ursodiol, actigall, ursofalk, ursolvan, delursan, ursodeoxycholate, urso, urso forte, ursodesoxycholic acid | |
CHEBI:9907 | |
CC(CCC(=O)O)C1CCC2C1(CCC3C2C(CC4C3(CCC(C4)O)C)O)C |
Specifications
Ursodeoxycholic acid | |
Glass bottle | |
+58° to +62° (20°C, 589nm) (c=2, ethanol) | |
200°C to 206°C | |
128-13-2 | |
C24H40O4 | |
15, 10074 | |
RUDATBOHQWOJDD-UZVSRGJWSA-N | |
(4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid | |
31401 | |
392.57 | |
≥98.5% (Acid) |
Authentic | |
+60.00 (20.00°C c=2,C2H5OH) | |
Colorless | |
1g | |
99% | |
MFCD00003680 | |
ursodeoxycholic acid, ursodiol, actigall, ursofalk, ursolvan, delursan, ursodeoxycholate, urso, urso forte, ursodesoxycholic acid | |
CC(CCC(=O)O)C1CCC2C1(CCC3C2C(CC4C3(CCC(C4)O)C)O)C | |
392.58 | |
CHEBI:9907 | |
Solution |