Learn More
Ursodeoxycholic acid, 99%, ACROS Organics™
Brand: Acros Organics 208590010
Code : 29
Additional Details : CAS Number : 128-13-2 Weight : 0.05100kg
Packaging | Glass bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
128-13-2 | |
392.58 | |
RUDATBOHQWOJDD-UZVSRGJWSA-N | |
31401 | |
(4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid |
C24H40O4 | |
MFCD00003680 | |
ursodeoxycholic acid, ursodiol, actigall, ursofalk, ursolvan, delursan, ursodeoxycholate, urso, urso forte, ursodesoxycholic acid | |
CHEBI:9907 | |
CC(CCC(=O)O)C1CCC2C1(CCC3C2C(CC4C3(CCC(C4)O)C)O)C |
Specifications
Ursodeoxycholic acid | |
Glass bottle | |
+58° to +62° (20°C, 589nm) (c=2, ethanol) | |
200°C to 206°C | |
128-13-2 | |
C24H40O4 | |
15, 10074 | |
RUDATBOHQWOJDD-UZVSRGJWSA-N | |
(4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid | |
31401 | |
392.57 | |
≥98.5% (Acid) |
Authentic | |
+60.00 (20.00°C c=2,C2H5OH) | |
Colorless | |
1g | |
99% | |
MFCD00003680 | |
ursodeoxycholic acid, ursodiol, actigall, ursofalk, ursolvan, delursan, ursodeoxycholate, urso, urso forte, ursodesoxycholic acid | |
CC(CCC(=O)O)C1CCC2C1(CCC3C2C(CC4C3(CCC(C4)O)C)O)C | |
392.58 | |
CHEBI:9907 | |
Solution |