missing translation for 'onlineSavingsMsg'
Learn More

Urocanic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10553591
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
104-98-3
C6H6N2O2
138.13
MFCD00005203
LOIYMIARKYCTBW-UPHRSURJSA-N
urocanic acid, 4-imidazoleacrylic acid, trans-urocanic acid, urocaninic acid, imidazoleacrylic acid, 5-imidazoleacrylic acid, urocanate, imidazole-4-acrylic acid, 2e-3-1h-imidazol-4-yl acrylic acid, 3-1h-imidazol-4-yl-2-propenoic acid
1549103
CHEBI:30818
(Z)-3-(1H-imidazol-5-yl)prop-2-enoic acid
C1=C(NC=N1)C=CC(=O)O
This item is not returnable. View return policy
104-98-3
C6H6N2O2
138.13
MFCD00005203
LOIYMIARKYCTBW-UPHRSURJSA-N
urocanic acid, 4-imidazoleacrylic acid, trans-urocanic acid, urocaninic acid, imidazoleacrylic acid, 5-imidazoleacrylic acid, urocanate, imidazole-4-acrylic acid, 2e-3-1h-imidazol-4-yl acrylic acid, 3-1h-imidazol-4-yl-2-propenoic acid
1549103
CHEBI:30818
(Z)-3-(1H-imidazol-5-yl)prop-2-enoic acid
C1=C(NC=N1)C=CC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 217.0°C to 220.0°C
Color White to Beige
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 25, 126
Merck Index 10, 9691
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone,,insoluble in ethanol,diethyl ether
Formula Weight 138.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Urocanic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'