missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uridine, ∽99%, MP Biomedicals™
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Specifications
Color | White |
Assay Percent Range | ∽99% |
Quantity | 5 g |
Formula Weight | 244.2 |
Physical Form | Powder |
Chemical Name or Material | Uridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'