missing translation for 'onlineSavingsMsg'
Learn More

Uridine, 99%, ACROS Organics™

Brand:  Acros Organics 140775000

Code : 29

Additional Details : CAS Number : 58-96-8 Weight : 0.55000kg

Packaging Glass bottle
Quantity 500g

 View more versions of this product

Product Code. 10012783

  • £919.00 / 500g

In stock, ready to ship: 8

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

58-96-8
244.2
DRTQHJPVMGBUCF-DHRBYNEYSA-N
89495556
C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O
C9H12N2O6
MFCD00006526
uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd
1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione

Specifications

0.72 to 0.76 (A250/A260), 0.34 to 0.38 (A280/A260)
Authentic
Glass bottle
98% min. (lambda max. 262nm,pH 7) molar absorptivity 10100+/-2%,on dry basis
95% min. (c=1, H2O, 430nm) 1cm cell
162.0°C to 170.0°C
58-96-8
C9H12N2O6
31, 23
uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd
C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O
244.2
244.2
99%
Uridine
1% max. (105°C, 2 hrs)
+ 8.40
0.1% max.
White
500g
98.5% min. (HPLC)
MFCD00006526
15, 10062
DRTQHJPVMGBUCF-DHRBYNEYSA-N
1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
89495556
Crystalline Powder
Videos
SDS Documents
Certificates