missing translation for 'onlineSavingsMsg'
Learn More

Uric acid, +99%, ACROS Organics™

Brand:  Acros Organics 171291000

Code : 29

Additional Details : CAS Number : 69-93-2 Weight : 0.15000kg

Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10327400

  • £78.30 / 100g


EU Stock 8
Add to basket

Specifications

Chemical Identifiers

Uric acid
69-93-2
99+%
168.11
2, 6, 8-Trioxypurine
99+%
≥99.0% (ex N)
C5H4N4O3
MFCD00005712

Specifications

Uric acid
69-93-2
≥99.0% (ex N)
Authentic
0.5% max. (105°C, 3 hrs)
MFCD00005712
2, 6, 8-Trioxypurine
White to off-white
99+%
Fine Powder
99+%
C5H4N4O3
168.11
Plastic bottle
0.2% max.
100g
Videos
SDS