missing translation for 'onlineSavingsMsg'
Learn More

Urease, Merck Millipore

Lyophilized 5 U/mg EC 3.5.1.5

Brand:  Merck Millipore 1084890025

Code : NEW

Additional Details : CAS Number : 9002-13-5 Weight : 0.02500kg

Quantity 25 g

 View more versions of this product

Product Code. 10138370

  • £648.00 / 25g

Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Activity: ≥ 5 U/mg
  • Suitability for determination of urea: passes test
Specifications

Chemical Identifiers

6.0-7.0 (20°C, 5 g/L in H2O, slurry)

Specifications

25 g
6.0-7.0 (20°C, 5 g/L in H2O, slurry)
Videos
SDS
Documents
Certificates