missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urea, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
3 kg
Specifications
Melting Point | 131°C |
Color | White |
pH | 7.5 to 9.5 |
Packaging | Plastic powder jar |
Quantity | 1 kg |
Formula Weight | 60.06g/mol |
Vapor Pressure | 1.25mmHg 25 |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'