missing translation for 'onlineSavingsMsg'
Learn More

Urea (Colorless-to-White Crystals or Crystalline Powder/Mol. Biol.), Fisher BioReagents™

Catalog Number p-23675
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Poly Bottle
Poly Drum
Poly Pail
This item is not returnable. View return policy
57-13-6
CH4N2O
60.056
XSQUKJJJFZCRTK-UHFFFAOYSA-N
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
1176
CHEBI:48376
urea
C(=O)(N)N
This item is not returnable. View return policy
57-13-6
CH4N2O
60.056
XSQUKJJJFZCRTK-UHFFFAOYSA-N
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
1176
CHEBI:48376
urea
C(=O)(N)N
Urea is used as a denaturing agent for DNA and proteins.
TRUSTED_SUSTAINABILITY
Absorbance 0.05 max. (5M solution) at 280nm
Melting Point 131°C
Color White
pH 7.5 to 9.5
Assay Percent Range ≥99 %
Packaging Poly Bottle
Linear Formula NH2CONH2
Quantity 500 g
Merck Index 15, 10052
Formula Weight 60.06
Vapor Pressure 1.25mmHg at 25°C
Percent Purity ≥99%
Grade DNase-, RNase- and Protease-Free
Physical Form Solid
Chemical Name or Material Urea
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'