missing translation for 'onlineSavingsMsg'
Learn More

Urea, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number 15548274
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
57-13-6
CH4N2O
60.056
XSQUKJJJFZCRTK-UHFFFAOYSA-N
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
1176
CHEBI:48376
urea
C(=O)(N)N
This item is not returnable. View return policy
57-13-6
CH4N2O
60.056
XSQUKJJJFZCRTK-UHFFFAOYSA-N
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
1176
CHEBI:48376
urea
C(=O)(N)N

High quality chemicals for laboratory and specialized industrial use

Packaging Poly Bottle
Quantity 2.5 kg
Specific Gravity 1L = 1.32kg
Formula Weight 60.06
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Urea
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'