missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Idex Bottle Caps
Click to view available options
Thread Size (Metric):
GL 38
GL 45
Color:
Aqua Blue
Red
For Use With (Equipment):
0.06 in. O.D. tubing
Specifications
For Use With (Equipment) | 0.06 in. O.D. tubing |
Product Type | Cap |
Color | Aqua Blue |
Material | Polypropylene |
No. per Case | 1 Cs. |
Thread Size (Metric) | GL 38 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'