Learn More
Cytiva Untagged Column Package
Combines cation and anion exchange with size exclusion chromatography for purification of most untagged or native proteins
Brand: Cytiva 29497631
Additional Details : CAS Number : 64-17-5 Weight : 0.04500kg
Description
- Package contains HiTrap™ Capto Q ImpRes (1 x 1 mL) and Capto SP ImpRes (1 x 1 mL) Ion Exchange (IEX) columns for effective first-step purification
- Obtain final high purity with HiPrep™ 16/60 Sephacryl S-200 HR: a Size Exclusion Chromatography (SEC) column with a broad fractionation range of Mr 5000 to ∼ 250 000 for globular proteins
- Includes instructions with guidance and tips to quickly get started
Specifications
Untagged Column | |
1 Kit | |
4°C to 30°C |
Purification of Most Untagged or Native Proteins | |
HiTrap&Trade; Capto Q ImpRes (1 x 1 mL) Column, Capto SP ImpRes (1 x 1 mL) Ion Exchange (IEX) Column, and HiPrep™ 16/60 Sephacryl S-200 HR Size Exclusion Chromatography Column |
Safety and Handling
- Untagged package
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.