missing translation for 'onlineSavingsMsg'
Learn More

unsym-Dimethylethylenediamine, 99%, Thermo Scientific Chemicals

Catalog Number p-3736317
18.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
108-00-9
C4H12N2
88.15
MFCD00008175
DILRJUIACXKSQE-UHFFFAOYSA-N
n,n-dimethylethylenediamine, 2-dimethylaminoethylamine, 2-aminoethyl dimethylamine, 1,2-ethanediamine, n,n-dimethyl, 2-dimethylamino ethylamine, n,n-dimethyl-1,2-ethanediamine, n1,n1-dimethylethane-1,2-diamine, n,n-dimethyl-1,2-ethylenediamine, 2-aminoethyldimethylamine, n,n-dimethylethanediamine
66053
N',N'-dimethylethane-1,2-diamine
CN(C)CCN
This item is not returnable. View return policy
108-00-9
C4H12N2
88.15
MFCD00008175
DILRJUIACXKSQE-UHFFFAOYSA-N
n,n-dimethylethylenediamine, 2-dimethylaminoethylamine, 2-aminoethyl dimethylamine, 1,2-ethanediamine, n,n-dimethyl, 2-dimethylamino ethylamine, n,n-dimethyl-1,2-ethanediamine, n1,n1-dimethylethane-1,2-diamine, n,n-dimethyl-1,2-ethylenediamine, 2-aminoethyldimethylamine, n,n-dimethylethanediamine
66053
N',N'-dimethylethane-1,2-diamine
CN(C)CCN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -70°C
Color Colorless to Yellow
Density 0.8000g/mL
Boiling Point 105.5°C to 107°C
Flash Point 12°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NCH2CH2NH2
Refractive Index 1.4255 to 1.4275
Quantity 25 g
Beilstein 04, II, 690
Specific Gravity 0.8
Solubility Information Solubility in water: miscible with water
Formula Weight 88.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material unsym-Dimethylethylenediamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • unsym-Dimethylethylenediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'