Learn More
Superfos™ UniPak™ Polypropylene Bucket
Provides flexible packaging. Superfos™ UniPak™ Polypropylene Buckets match each lid diameter so only one versatile type of lid is needed for a range of sizes. Tamper resistant.
£0.06 - £0.60
Specifications
Material Polypropylene
Autoclavable Yes
Lid Standard lid, membrane lid
Certifications/Compliance European Union Commission Regulation No. 10/2011/EC with amendments
View More Specs
Products 7
Product Code Brand Capacity (Metric) Handle Material Color Price Quantity & Availability  
Product Code Brand Capacity (Metric) Handle Material Color Price Quantity & Availability  
10723647 View accessory products View Documents Superfos™
513319904
1.6L Flexible Plastic Translucent £0.60
Each View alternative products
Save Up to 
»
Estimated Shipment
13-01-2020
10129994 View Documents Superfos™
5030-WHITE
1000mL Flexible Plastic White £0.43
Each View alternative products
Save Up to 
»
Estimated Shipment
13-01-2020
10575195 View accessory products View Documents Superfos™
5031
1.18L Flexible Plastic White £0.38
Each View alternative products
Save Up to 
»
Estimated Shipment
13-01-2020
10499204 View accessory products View Documents Superfos™
5011-WHITE
280mL N/A White £0.06
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
10725356 View accessory products View Documents Superfos™
5110
365mL Flexible Plastic Translucent £0.19
Each View alternative products
Save Up to 
»
Estimated Shipment
13-01-2020
10419214 View accessory products View Documents Superfos™
5012-WHITE
520mL N/A White £0.20
Each View alternative products
Save Up to 
»
Estimated Shipment
13-01-2020
10626185 View accessory products View Documents Superfos™
5022-WHITE
870mL Flexible Plastic White £0.34
Each View alternative products
Save Up to 
»
Estimated Shipment
13-01-2020
Description
Description
  • Protective collar
  • Withstands hot filling and blast freezing
  • Available in round, rectangular, oval and square shapes
  • Broad range of standard sizes
  • Liquid tight, snap-on lid
  • Same lid fits several volumes
  • Excellent decoration options with In-Mould Labelling
  • Visible deterrent to tampering
Specifications
Specifications
Polypropylene
Yes
Standard lid, membrane lid
European Union Commission Regulation No. 10/2011/EC with amendments