Learn More
Kartell™ Unequal Straight Connectors with Tapered Ends
Created from Polypropylene. Kartell™ Unequal Straight Connectors with Tapered Ends features and inlet and an outlet with different diameters.
Brand: Kartell™ 0087900
Code : A0
Additional Details : Weight : 0.01000kg
Description
Created from Polypropylene. Kartell™ Unequal Straight Connectors with Tapered Ends features and inlet and an outlet with different diameters.
Polypropylene
Inlet and out have different diameters
CE and IVD Certifications
Tapered Ends
Specifications
Polypropylene | |
Straight Tapered Connectors | |
4.6/7.5mm |
8,10,12,14,16 mm | |
10 pieces |