missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Undecanoic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
Undecanoic acid is used as an unsaturated fatty acid. It is also used as intermediates of Liquid Crystals. It is used in organic synthesis, Catalytic agent, Petrochemical additive. It is also used in anti-fungal agent, perfumery chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Undecanoic acid is used as an unsaturated fatty acid. It is also used as intermediates of Liquid Crystals. It is used in organic synthesis, Catalytic agent, Petrochemical additive. It is also used in anti-fungal agent, perfumery chemicals.
Solubility
Insoluble in water.
Notes
Store at 4°C. Incompatible with bases, oxidizing agents, reducing agents. Keep container closed when not in use.
Undecanoic acid is used as an unsaturated fatty acid. It is also used as intermediates of Liquid Crystals. It is used in organic synthesis, Catalytic agent, Petrochemical additive. It is also used in anti-fungal agent, perfumery chemicals.
Solubility
Insoluble in water.
Notes
Store at 4°C. Incompatible with bases, oxidizing agents, reducing agents. Keep container closed when not in use.
Specifications
Melting Point | 26°C to 29°C |
Boiling Point | 283°C to 285°C |
Flash Point | >110°C (230°F) |
Quantity | 50 g |
Beilstein | 1759287 |
Solubility Information | Insoluble in water. |
Formula Weight | 186.3 |
Percent Purity | 98% |
Chemical Name or Material | Undecanoic acid |
Safety and Handling
Signal Word
- Warning
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'