missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC5H2/UNC5B Mouse anti-Rat, Clone: 187305, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | UNC5H2/UNC5B |
---|---|
Clone | 187305 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant rat UNC5H2 Gly27-Asp373 Accession # O08722 |
Gene ID (Entrez) | 219699 |
Gene Accession No. | O08722 |
Description
UNC5H2/UNC5B Monoclonal specifically detects UNC5H2/UNC5B in Rat samples. It is validated for Western Blot.Specifications
UNC5H2/UNC5B | |
Mouse myeloma cell line NS0-derived recombinant rat UNC5H2 Gly27-Asp373 Accession # O08722 | |
O08722 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
187305 | |
219699 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects rat UNC5H2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant rat UNC5H1 or recombinant human UNC5H4 is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only