missing translation for 'onlineSavingsMsg'
Learn More

UNC 926 hydrochloride, Tocris Bioscience™

L3MBTL1 domain inhibitor

Brand:  RD Systems (Bio-Techne) 4516/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13464864

  • £135.00 / 10mg


Estimated Shipment
03-04-2019
Add to basket

Description

Description

Methyl-lysine (Kme) reader domain inhibitor; binds to the MBT domain of the L3MBTL1 protein (K d = 3.9 μM). Selectively inhibits the L3MBTL1 3XMBT -H4K20me1 interaction in a peptide pull down assay.
Specifications

Chemical Identifiers

1184136-10-4
373.719
unc 926 hydrochloride, unc-926 hydrochloride, 3-bromophenyl 4-1-prrolidinyl-1-piperidinyl methanone hydrochloride
(3-bromophenyl)-(4-pyrrolidin-1-ylpiperidin-1-yl)methanone;hydrochloride
C16H22BrClN2O
DPPMBBHOXQOMJI-UHFFFAOYSA-N
90488947
C1CCN(C1)C2CCN(CC2)C(=O)C3=CC(=CC=C3)Br.Cl

Specifications

UNC 926 hydrochloride
1184136-10-4
unc 926 hydrochloride, unc-926 hydrochloride, 3-bromophenyl 4-1-prrolidinyl-1-piperidinyl methanone hydrochloride
C1CCN(C1)C2CCN(CC2)C(=O)C3=CC(=CC=C3)Br.Cl
373.719
373.72
10mg
C16H22BrClN2O
DPPMBBHOXQOMJI-UHFFFAOYSA-N
(3-bromophenyl)-(4-pyrrolidin-1-ylpiperidin-1-yl)methanone;hydrochloride
90488947
>99%
Videos