missing translation for 'onlineSavingsMsg'
Learn More

UNC 0224, Tocris Bioscience™

Potent G9a and GLP inhibitor

Brand:  RD Systems (Bio-Techne) 3861/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13433714

  • £163.17 / 10mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent G9a and GLP inhibitor (IC50 values are 15 and 20 nM in a ThioGlo assay, respectively). Exhibits >1000-fold selectivity for G9a over SET7, SET8 and SET9.
Specifications

Chemical Identifiers

1197196-48-7
485.677
7-3-dimethylamino propoxy-6-methoxy-2-4-methyl-1,4-diazepan-1-yl-n-1-methylpiperidin-4-yl quinazolin-4-amine, n-debenzyl-n-methyl-o-demethyl-o-3-dimethylaminopropyl-bix 01294, 7-3-dimethylamino propoxy-2-hexahydro-4-methyl-1h-1,4-diazepin-1-yl-6-methoxy-n-1-methyl-4-piperidinyl-4-quinazolinamine
7-[3-(dimethylamino)propoxy]-6-methoxy-2-(4-methyl-1,4-diazepan-1-yl)-N-(1-methylpiperidin-4-yl)quinazolin-4-amine
C26H43N7O2
XIVUGRBSBIXXJE-UHFFFAOYSA-N
44251522
CN1CCCN(CC1)C2=NC3=CC(=C(C=C3C(=N2)NC4CCN(CC4)C)OC)OCCCN(C)C

Specifications

10mg
1197196-48-7
7-3-dimethylamino propoxy-6-methoxy-2-4-methyl-1,4-diazepan-1-yl-n-1-methylpiperidin-4-yl quinazolin-4-amine, n-debenzyl-n-methyl-o-demethyl-o-3-dimethylaminopropyl-bix 01294, 7-3-dimethylamino propoxy-2-hexahydro-4-methyl-1h-1,4-diazepin-1-yl-6-methoxy-n-1-methyl-4-piperidinyl-4-quinazolinamine
CN1CCCN(CC1)C2=NC3=CC(=C(C=C3C(=N2)NC4CCN(CC4)C)OC)OCCCN(C)C
485.677
485.67
UNC 0224
C26H43N7O2
XIVUGRBSBIXXJE-UHFFFAOYSA-N
7-[3-(dimethylamino)propoxy]-6-methoxy-2-(4-methyl-1,4-diazepan-1-yl)-N-(1-methylpiperidin-4-yl)quinazolin-4-amine
44251522
>98%
Videos
Documents