Learn More
UNC 0224, Tocris Bioscience™
Potent G9a and GLP inhibitor
POA
Description
Description
Potent G9a and GLP inhibitor (IC50 values are 15 and 20 nM in a ThioGlo assay, respectively). Exhibits >1000-fold selectivity for G9a over SET7, SET8 and SET9.