Learn More
Thermo Scientific™ Unassembled Convenience Kits: EPA Vials with Caps and Septa
Designed for discrete water sampling applications.
£85.10 - £103.02
Product Code | Brand | Volume (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Color | Price | Quantity & Availability | ||||
12478636
|
Thermo Scientific™
B7920-B |
20 mL | Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12428646
|
Thermo Scientific™
B7921-B |
20 mL | Amber |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12438656
|
Thermo Scientific™
B7951-B |
40 mL | Amber |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.
Each carton contains shrink-wrapped vials and separately packaged caps and natural-colored PTFE/clear Silicone septa in polybags.Specifications
Discrete Water Sampling Applications |