Hielscher Ultrasonics™ Ultrasonics Sonotrode


Overview
Brand: Hielscher Ultrasonics™

Manufacturer Part Number: S7

Sonotrode S7 aus Titan, Ø 7 mm, Länge ca. 100 mm,für Proben

UNSPSC: 41101518

Code: 50

Additional Details:
Additional Details: Weight: 0.96600kg



Product Code. 10093964

Quantity Price
1 £ 413.48 / Each
Estimated Shipment
07-04-2017
Add to basket