Learn More
Fisherbrand™ Model 705 Sonic Dismembrator Accessory
4 Tip horn for use with 500 or 700 watt sonicator systems; replacement tip also available.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 2.50000kg
Description
- For processing 0.5 to 15ml volumes
- Process 4 samples at one time
Specifications
4-tip Horn | |
Lyse cells, shear DNA, dissolve solids | |
FB705 Sonic Dismembrator and Q700 Sonicator |