Learn More
Excel Healthcare Ltd™ Ultrasonic Detergent
Efficient Cleaning Solution
Brand:  Excel Healthcare Ltd™ 10902
Additional Details : Weight : 5.00000kg
Product Code. 16310122
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Ultrasonic Cleaning Enhances The Removal Of Debris Prior To Sterilisation
  • It Is Vital That The Water And Detergent Are Well Maintained, Cleaned And Changed At Regular Intervals To Ensure Maximum Cleaning Effectiveness
  • Htm 01-05 Recommends That Intruments Placed In Ultrasonic Baskets Must Be Fully Immersed In The Cleaning Solution
  • After The Process Has Taken Place, Instruments Must Be Thoroughly Cleaned And Rinsed Using Distilled Water In Order To Remove Residual Soil And Detergents With Minimum Risk Of Salt Deposition
Specifications
Specifications
1
500 mL
Double headed bottle
Detergent for Ultrasonic Cleaners
Ultrasonic Cleaner