Learn More
Corning™ UltraGAPS™ Microarray Slides
Ideal for printing both long (>50 mer) oligonucleotide and cDNA arrays
Brand:  Corning™ 40017
Code : 67
Additional Details : Weight : 0.22680kg
 View more versions of this product
Product Code. 10164112
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
  • Uniform, covalently bound coating of pure Gamma Amino Propyl Silane (GAPS) yields a high-quality DNA-binding surface with stable surface properties and low autofluorescence
  • Hydrophobic and stable surface yields smaller spots and increased slide-to-slide reproducibility
  • Low background, high signal-to-noise ratio, and consistent spot morphology enhance the ability to detect the expression of low abundance transcripts
  • Cleaned and individually examined for mechanical defects and the presence of glass and dust particles
  • Shipped with a lot-specific Quality Certificate, detailing the results of nine quality parameters, including DNA retention and spot morphology
Specifications
Specifications
Yes
UltraGAPS™
25/Pk.
75.5mm
No
Microarray Slide
For printing both long (>50 mer) oligonucleotide and cDNA arrays
25.3mm
1.1mm
Videos
Documentation