missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ UltraComp eBeads

Designed for use in compensation with all fluorochromes excited by ultraviolet, violet, blue, green, yellow-green, and red lasers. UltraComp eBeads™ react with antibodies of mouse, rat, and hamster origin, and are immunoglobulin light chain-independent. 1 drop (50μL)/test.

Brand:  Invitrogen™ 01-2222-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15536296

  • £72.36 / 25 tests


EU Stock 14
Add to basket

Alert:

Contains Sodium azide

Disclaimers:

For Research Use Only


Description

Description

  • Each drop of beads contains both a positive population that will capture any mouse, rat or hamster antibody and a negative population that will not react with antibody
  • This bimodal distribution can be used for single-color compensation controls in multicolor flow cytometry experiments
  • Compatible with all fluorochromes excited by an ultraviolet (355nm) or violet (405nm) laser
  • Should be used with standard staining buffers which contain PBS or HBSS, protein such as BSA or FBS, and sodium azide. No other additives should be used.
Specifications

Specifications

Flow Cytometric Analysis
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
25 tests
2° to 8°C. Do not freeze.
Videos