missing translation for 'onlineSavingsMsg'
Learn More

UK 5099, Tocris Bioscience™

MCT inhibitor; also inhibits of pyruvate transport

Brand:  Tocris (Bio-Techne) 418610

Code : Z1

Additional Details : Weight : 0.01100kg

Quantity 10mg

 View more versions of this product

Product Code. 13464304

  • £155.76 / 10mg


Estimated Shipment
29-03-2019
Add to basket

Description

Description

Inhibitor of plasma membrane monocarboxylate transporters (MCTs) and the mitochondrial pyruvate carrier (MPC).
Specifications

Chemical Identifiers

56396-35-1
287.298
2-cyano-3-1-phenyl-1h-indol-3-yl-2-propenoic acid, 2e-2-cyano-3-1-phenylindol-3-yl prop-2-enoate
(E)-2-cyano-3-(1-phenylindol-3-yl)prop-2-enoate
C18H11N2O2-
BIZNHCWFGNKBBZ-JLHYYAGUSA-M
71772548
C1=CC=C(C=C1)N2C=C(C3=CC=CC=C32)C=C(C#N)C(=O)[O-]

Specifications

UK 5099
56396-35-1
2-cyano-3-1-phenyl-1h-indol-3-yl-2-propenoic acid, 2e-2-cyano-3-1-phenylindol-3-yl prop-2-enoate
C1=CC=C(C=C1)N2C=C(C3=CC=CC=C32)C=C(C#N)C(=O)[O-]
287.298
288.3
10mg
C18H11N2O2-
BIZNHCWFGNKBBZ-JLHYYAGUSA-M
(E)-2-cyano-3-(1-phenylindol-3-yl)prop-2-enoate
71772548
>99%
Videos