missing translation for 'onlineSavingsMsg'
Learn More

UK 14,304, Tocris Bioscience™

α2 agonist

Brand:  Tocris (Bio-Techne) 0425/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13487633

  • £114.75 / 10mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Full α2 adrenergic agonist. Centrally active following systemic administration in vivo. Also available as part of the α2-Adrenoceptor Tocriset™ and Mixed Adrenergic Tocriset™ . Water soluble salt version UK 14,304 tartrate (Cat. No. 2466) also available.
Specifications

Chemical Identifiers

59803-98-4
292.14
5-Bromo-6-(2-imidazolin-2-ylamino)quinoxaline
CHEBI:3175
C1CN=C(N1)NC2=C(C3=NC=CN=C3C=C2)Br
C11H10BrN5
XYLJNLCSTIOKRM-UHFFFAOYSA-N
2435
5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine

Specifications

UK 14, 304
10mg
C11H10BrN5
XYLJNLCSTIOKRM-UHFFFAOYSA-N
5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine
2435
292.14
59803-98-4
>98%
5-Bromo-6-(2-imidazolin-2-ylamino)quinoxaline
C1CN=C(N1)NC2=C(C3=NC=CN=C3C=C2)Br
292.14
CHEBI:3175
Videos