missing translation for 'onlineSavingsMsg'
Learn More

UK 14,304 tartrate, Tocris Bioscience™

α2 agonist. Water-soluble form of UK 14,304 (Cat. No. 0425)

Brand:  Tocris (Bio-Techne) 2466/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13461264

  • £132.30 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Water-soluble form of UK 14,304 (Cat. No. 0425). Full α2 adrenergic agonist. Centrally active following systemic administration in vivo.
Specifications

Chemical Identifiers

70359-46-5
442.226
brimonidine tartrate, --tartaric acid; brimonidine
5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine;(2R,3S)-2,3-dihydroxybutanedioic acid
C15H16BrN5O6
QZHBYNSSDLTCRG-YKWZRPGSSA-N
72376318
C1CN=C(N1)NC2=C(C3=NC=CN=C3C=C2)Br.C(C(C(=O)O)O)(C(=O)O)O

Specifications

UK 14, 304 tartrate
70359-46-5
brimonidine tartrate, --tartaric acid; brimonidine
C1CN=C(N1)NC2=C(C3=NC=CN=C3C=C2)Br.C(C(C(=O)O)O)(C(=O)O)O
442.226
442.23
10mg
C15H16BrN5O6
QZHBYNSSDLTCRG-YKWZRPGSSA-N
5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine;(2R,3S)-2,3-dihydroxybutanedioic acid
72376318
>99%
Videos