Learn More
(+)-UH 232 maleate, Tocris Bioscience™
D2 autoreceptor antagonist. Also D3 partial agonist
Brand:  Tocris (Bio-Techne) 077550
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13487983
  • £539.52 / 50mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
D2 antagonist (Ki = 72.7 nM in a ligand binding assay; apparent KB = 14.5 nM in a cAMP accumulation assay); displays preferential activity at central dopamine autoreceptors. Stimulates a marked acceleration of dopamine synthesis and turnover. Produces locomotor stimulation. Exhibits little or no activity at central noradrenalin and 5-HT receptors. Also D3 partial agonist.
Specifications
Chemical Identifiers
1217473-50-1
391.508
cis-(+)-5-Methoxy-1-methyl-2-(di-N-propylamino)tetralin maleate
C22H33NO5
MEQAJDYHKYAPJE-GUUGRXDUSA-N
46830250
Specifications
(+)-UH 232 maleate
1217473-50-1
C22H33NO5
MEQAJDYHKYAPJE-GUUGRXDUSA-N
(Z)-but-2-enedioic acid;(1S,2R)-5-methoxy-1-methyl-N,N-dipropyl-1,2,3,4-tetrahydronaphthalen-2-amine
46830250
50mg
>98%
cis-(+)-5-Methoxy-1-methyl-2-(di-N-propylamino)tetralin maleate
CCCN(CCC)C1CCC2=C(C1C)C=CC=C2OC.C(=CC(=O)O)C(=O)O
391.508
391.51
Videos