Learn More
UCL 2077, Tocris Bioscience™
Slow afterhyperpolarization (sAHP) channel blocker; sub-type selective Kv channel effects
Brand:  Tocris (Bio-Techne) 266110
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13461674
  • £139.65 / 10mg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
Slow afterhyperpolarization (sAHP) channel blocker; reduces sAHP in hippocampal slice preparations. Displays no effect on Ca2+ currents or the time course of sAHP/sIAHP. Exhibits potent inhibition of KCNQ1 and KCNQ2, but differential effects at other KCNQs. Rescues memory retrieval in dopamine β-hydroxylase knockout mice.
Specifications
Chemical Identifiers
918311-87-2
350.465
3-triphenylmethylaminomethyl pyridine, pyridin-3-ylmethyl triphenylmethyl amine, n-trityl-3-pyridinemethanamine, 3-tribenzylaminomethyl pyridine, ucl hplc , solid, 1,1,1-triphenyl-n-pyridin-3-yl methyl methanamine
1,1,1-triphenyl-N-(pyridin-3-ylmethyl)methanamine
C25H22N2
PQFNWDHABGBCHB-UHFFFAOYSA-N
24868317
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NCC4=CN=CC=C4
Specifications
10mg
918311-87-2
3-triphenylmethylaminomethyl pyridine, pyridin-3-ylmethyl triphenylmethyl amine, n-trityl-3-pyridinemethanamine, 3-tribenzylaminomethyl pyridine, ucl hplc , solid, 1,1,1-triphenyl-n-pyridin-3-yl methyl methanamine
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NCC4=CN=CC=C4
350.465
350.46
UCL 2077
C25H22N2
PQFNWDHABGBCHB-UHFFFAOYSA-N
1,1,1-triphenyl-N-(pyridin-3-ylmethyl)methanamine
24868317
>99%