missing translation for 'onlineSavingsMsg'
Learn More

UCB 35625, Tocris Bioscience™

Potent CCR1 and CCR3 antagonist

Brand:  Tocris (Bio-Techne) 275710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13430534

  • £289.59 / 10mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent chemokine CCR1 and CCR3 receptor antagonist. Inhibits MIP-1α-induced chemotaxis in CCR1 transfectants and eotaxin-induced chemotaxis in CCR3 transfectants (IC50 values are 9.57 and 93.8 nM respectively). Antagonizes CCR3-mediated entry of HIV-1 isolate 89.6 into NP-2 cells (IC50 = 57 nM).

Isomer also available.

Specifications

Chemical Identifiers

301648-08-8
655.442
unii-hd099hot7h, unii-h942hgf14d, hd099hot7h, chembl34626, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide 1:1 , cis, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide 1:1 , trans, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide, cis, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide, trans, unii-e32u2esr7j component foafbmysxigaox-kekkvsgzsa-n, unii-e32u2esr7j component foafbmysxigaox-mumzcososa-n
2,7-dichloro-N-[1-[[(1E)-cycloocten-1-yl]methyl]-1-ethylpiperidin-1-ium-4-yl]-9H-xanthene-9-carboxamide;iodide
C30H37Cl2IN2O2
FOAFBMYSXIGAOX-LQGGPMKRSA-N
6918496
CC[N+]1(CCC(CC1)NC(=O)C2C3=C(C=CC(=C3)Cl)OC4=C2C=C(C=C4)Cl)CC5=CCCCCCC5.[I-]

Specifications

UCB 35625
301648-08-8
unii-hd099hot7h, unii-h942hgf14d, hd099hot7h, chembl34626, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide 1:1 , cis, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide 1:1 , trans, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide, cis, piperidinium, 1-1-cycloocten-1-ylmethyl-4-2,7-dichloro-9h-xanthen-9-yl carbonyl amino-1-ethyl-, iodide, trans, unii-e32u2esr7j component foafbmysxigaox-kekkvsgzsa-n, unii-e32u2esr7j component foafbmysxigaox-mumzcososa-n
CC[N+]1(CCC(CC1)NC(=O)C2C3=C(C=CC(=C3)Cl)OC4=C2C=C(C=C4)Cl)CC5=CCCCCCC5.[I-]
655.442
655.44
10mg
C30H37Cl2IN2O2
FOAFBMYSXIGAOX-LQGGPMKRSA-N
2,7-dichloro-N-[1-[[(1E)-cycloocten-1-yl]methyl]-1-ethylpiperidin-1-ium-4-yl]-9H-xanthene-9-carboxamide;iodide
6918496
>98%
Videos
Documents