Learn More
UBP1112, Tocris Bioscience™
Group III mGlu antagonist
Brand:  Tocris (Bio-Techne) 136950
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13439243
  • £520.38 / 50mg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
A selective group III mGlu receptor antagonist (apparent Kd values are 5.1 and 488 μM for group III and group II mGlu receptors respectively; IC50 > 1 mM for group I, NMDA, AMPA and kainate receptors).
Specifications
Chemical Identifiers
339526-74-8
259.198
2-amino-2-3-methyl-4-phosphonophenyl propanoic acid
2-amino-2-(3-methyl-4-phosphonophenyl)propanoic acid
C10H14NO5P
LLEOLNUKLKCAFG-UHFFFAOYSA-N
5022489
CC1=C(C=CC(=C1)C(C)(C(=O)O)N)P(=O)(O)O
Specifications
UBP1112
C10H14NO5P
LLEOLNUKLKCAFG-UHFFFAOYSA-N
2-amino-2-(3-methyl-4-phosphonophenyl)propanoic acid
5022489
>99%
339526-74-8
2-amino-2-3-methyl-4-phosphonophenyl propanoic acid
CC1=C(C=CC(=C1)C(C)(C(=O)O)N)P(=O)(O)O
259.198
259.2