missing translation for 'onlineSavingsMsg'
Learn More

UBP 282, Tocris Bioscience™

AMPA/kainate antagonist; distinguishes between motoneuron and dorsal root kainate receptors

Brand:  Tocris (Bio-Techne) 176550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13459953

  • £460.22 / 50mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

AMPA and kainate receptor antagonist. Inhibits AMPA receptor-, but not kainate receptor-mediated currents on spinal neonatal motoneurons yet antagonizes kainate-induced responses on dorsal root C-fibres.
Specifications

Chemical Identifiers

544697-47-4
333.3
vbp, tocris-1765, d02sjn, 3-cbw, vbp series muscular dystrophy/asthma, vbp series muscular dystrophy/asthma , validus biopharma, s-1-2-amino-2-carboxyethyl-3-4-carboxybenzyl pyrimidine-2,4-dione, 4-3-2s-2-amino-2-carboxyethyl-2,6-dioxopyrimidin-1-yl methyl benzoic acid, 4-3-2s-2-amino-3-hydroxy-3-oxopropyl-2,6-dioxopyrimidin-1-yl methyl benzoic acid, ?s-?-amino-3-4-carboxyphenyl methyl-3,4-dihydro-2,4-dioxo-1 2h-pyrimidinepropanoic acid
4-[[3-[(2S)-2-amino-2-carboxyethyl]-2,6-dioxopyrimidin-1-yl]methyl]benzoic acid
C15H15N3O6
XLRLZPOBHPIDFX-NSHDSACASA-N
6604912
C1=CC(=CC=C1CN2C(=O)C=CN(C2=O)CC(C(=O)O)N)C(=O)O

Specifications

UBP 282
544697-47-4
vbp, tocris-1765, d02sjn, 3-cbw, vbp series muscular dystrophy/asthma, vbp series muscular dystrophy/asthma , validus biopharma, s-1-2-amino-2-carboxyethyl-3-4-carboxybenzyl pyrimidine-2,4-dione, 4-3-2s-2-amino-2-carboxyethyl-2,6-dioxopyrimidin-1-yl methyl benzoic acid, 4-3-2s-2-amino-3-hydroxy-3-oxopropyl-2,6-dioxopyrimidin-1-yl methyl benzoic acid, ?s-?-amino-3-4-carboxyphenyl methyl-3,4-dihydro-2,4-dioxo-1 2h-pyrimidinepropanoic acid
C1=CC(=CC=C1CN2C(=O)C=CN(C2=O)CC(C(=O)O)N)C(=O)O
333.3
333.3
50mg
C15H15N3O6
XLRLZPOBHPIDFX-NSHDSACASA-N
4-[[3-[(2S)-2-amino-2-carboxyethyl]-2,6-dioxopyrimidin-1-yl]methyl]benzoic acid
6604912
>98%
Videos