missing translation for 'onlineSavingsMsg'
Learn More

Ub-K63, clone: HWA4C4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-6077-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15297697

  • £157.28 / 100µg


Estimated Shipment
31-07-2018
Add to basket

Description

Description

Description: Ubiquitin is a highly conserved 76kDa protein which is ligated to many proteins in both monomeric and polymeric forms. Ubiquitin polymers (polyubiquitin) can be found in several varieties depending on which of its 7 lysine residues is the site of ligation. The two most common polymer chains are linked via an isopeptide bond between glycine-76 and either lysine-48 (K-48) or lysine-63 (K-63). K-48-linked polyubiquitination typically results in targeting the protein to the 26S proteosome resulting in proteolysis, although recently exceptions to this have been observed. In contrast, K-63-linked polyubiquitination results in signals related to intracellular trafficking, cell signaling, ribosomal biogenesis, and DNA damage repair. There are also reports that K63-linked polyubiquitin is involved in proteosome-independent proteolysis via autophagy.HWA4C4 reacts specifically with the isopeptide linkage region of lysine-63-linked polyubiquitin. It does not react with other forms of ubiquitin including ubiquitin monomer and lysine-48-linked chains.Applications Reported: This HWA4C4 antibody has been reported for use in immunoblotting (WB).Applications Tested: This HWA2-8°C4 antibody has been tested by immunoblot of lysine-63-linked polyubiquitin chains. This can be used at less than or equal to 2 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Ub-K63
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P0CG47
K63-linked polyubiquitin, Lysine63-linked poly ubiquitin chain
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
HWA4C4
Unconjugated
UBB
Purified
UBB
IgG2a
100μg
Store at 2-8°C.
Primary
7314
Videos

For Research Use Only.