missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand Fisherbrand U.S. Standard Stainless Steel Sieves, 8 in. dia. x 2 in.D

Brand:  Fisherbrand

Fisherbrand U.S. Standard Stainless Steel Sieves, 8 in. dia. x 2 in.D

Code : NEW

Additional Details : Weight : 0.09071kg

Product Code. 15879494

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.