missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ U-73343, 98%

Inactive analog of U-73122

Brand:  Alfa Aesar™ J62719.MB

Code : NEW

Additional Details : CAS Number : 142878-12-4 Weight : 0.00025kg

 View more versions of this product

Product Code. 15495029

  • POA / 25mg

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Specifications

25mg
Powder
0.98
C29H42N2O3
1-[6-[((17beta)-3-Methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]pyrrolidine-2,5-dione
Signal Transduction Reagent
White to Off-white
Inactive analog of U-73122
142878-12-4
466.66g/mol
136° to 138°C
Videos
SDS