Learn More
U-54494A hydrochloride, Tocris Bioscience™
κ agonist
Brand:  Tocris (Bio-Techne) 049850
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13487733
  • £557.72 / 50mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
κ-opioid agonist and anticonvulsant. Perhaps acts as an NMDA antagonist.
Specifications
Chemical Identifiers
112465-94-8
391.761
cis-(±)-3, 4-Dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzamide hydrochloride
C18H25Cl3N2O
WFUASZXAHZXJMX-UHFFFAOYSA-N
21276975
Specifications
U-54494A hydrochloride
112465-94-8
C18H25Cl3N2O
WFUASZXAHZXJMX-UHFFFAOYSA-N
3,4-dichloro-N-methyl-N-(2-pyrrolidin-1-ylcyclohexyl)benzamide;hydrochloride
21276975
50mg
>98%
cis-(±)-3, 4-Dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzamide hydrochloride
CN(C1CCCCC1N2CCCC2)C(=O)C3=CC(=C(C=C3)Cl)Cl.Cl
391.761
391.77
Videos