missing translation for 'onlineSavingsMsg'
Learn More

(-)-U-50488 hydrochloride, Tocris Bioscience™

Standard selective κ agonist. More active enantiomer of (±)-U-50488 (Cat. No. 0495)

Brand:  Tocris (Bio-Techne) 049650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13447733

  • £583.59 / 50mg

Estimated Shipment date
from Supplier 26-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

More active enantiomer of (±)-U-50488 .

Racemate and (+)-enantiomer also available.

Specifications

Chemical Identifiers

114528-79-9
423.803
--trans-1s,2s-u-50488 hydrochloride hydrate, trans-1s,2s-3,4-dichloro-n-methyl-n-2-1-pyrrolidinyl cyclohexyl benzeneacetamide hydrochloride hydrate, --trans-1s,2s-u-50488 hydrochloride hydrate, solid hplc, 2-3,4-dichlorophenyl-n-methyl-n-1s,2s-2-pyrrolidin-1-yl cyclohexyl acetamide hydrate hydrochloride
2-(3,4-dichlorophenyl)-N-methyl-N-[(1S,2S)-2-pyrrolidin-1-ylcyclohexyl]acetamide;hydrate;hydrochloride
C19H29Cl3N2O2
UWPRPWMHQDPOFR-MPGISEFESA-N
16220065
CN(C1CCCCC1N2CCCC2)C(=O)CC3=CC(=C(C=C3)Cl)Cl.O.Cl

Specifications

50mg
114528-79-9
--trans-1s,2s-u-50488 hydrochloride hydrate, trans-1s,2s-3,4-dichloro-n-methyl-n-2-1-pyrrolidinyl cyclohexyl benzeneacetamide hydrochloride hydrate, --trans-1s,2s-u-50488 hydrochloride hydrate, solid hplc, 2-3,4-dichlorophenyl-n-methyl-n-1s,2s-2-pyrrolidin-1-yl cyclohexyl acetamide hydrate hydrochloride
CN(C1CCCCC1N2CCCC2)C(=O)CC3=CC(=C(C=C3)Cl)Cl.O.Cl
423.803
405.79
(-)-U-50488 hydrochloride
C19H29Cl3N2O2
UWPRPWMHQDPOFR-MPGISEFESA-N
2-(3,4-dichlorophenyl)-N-methyl-N-[(1S,2S)-2-pyrrolidin-1-ylcyclohexyl]acetamide;hydrate;hydrochloride
16220065
>99%
Videos
Documents